Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A35 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SLC25A35 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC25A35 Polyclonal specifically detects SLC25A35 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SLC25A35 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
399512 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: IYMVKTHLQAQAASEIAVGHQYKHQGMFQALTEIGQKHGLVGLWRGALGGLPRVIVGSSTQLCTFSSTKDLLSQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin | |
Polyclonal | |
Rabbit | |
Human | |
FLJ40217, MGC120446, MGC120448, solute carrier family 25 member 35, solute carrier family 25, member 35 | |
SLC25A35 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title