Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A45 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15987620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15987620 20 μL
NBP159876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15987620 Supplier Novus Biologicals Supplier No. NBP15987620UL

Rabbit Polyclonal Antibody

SLC25A45 Polyclonal specifically detects SLC25A45 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC25A45
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8N413-2
Gene Alias solute carrier family 25 member 45, solute carrier family 25, member 45
Gene Symbols SLC25A45
Host Species Rabbit
Immunogen Synthetic peptides corresponding to LOC283130 The peptide sequence was selected from the C terminal of LOC283130. Peptide sequence GLRRRVYQGMLDCMVSSIRQEGLGVFFRGVTINSARAFPVNAVTFLSYEY.
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283130
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.