Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A48 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17951520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17951520 20 μL
NBP179515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17951520 Supplier Novus Biologicals Supplier No. NBP17951520UL

Rabbit Polyclonal Antibody

SLC25A48 Polyclonal specifically detects SLC25A48 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC25A48
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. BAC86705
Gene Alias FLJ44862, HCC-down-regulated mitochondrial carrier protein, HDMCP, solute carrier family 25 member 48, solute carrier family 25, member 48
Gene Symbols SLC25A48
Host Species Rabbit
Immunogen Synthetic peptide directed towards the c terminal of human LOC153328. Peptide sequence TAVGQLGNCHALLSPGGGQDTFRYSPNNSLLGTYSVPGPLPPQSHPFPMQ.
Molecular Weight of Antigen 31 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153328
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.