Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC26A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | SLC26A2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18052220
![]() |
Novus Biologicals
NBP18052220UL |
20 μL |
Each for $158.00
|
|
|||||
NBP180522
![]() |
Novus Biologicals
NBP180522 |
100 μL |
Each for $487.50
|
|
|||||
Description
SLC26A2 Polyclonal specifically detects SLC26A2 in Mouse samples. It is validated for Western Blot.Specifications
SLC26A2 | |
Polyclonal | |
Rabbit | |
NP_031911 | |
1836 | |
Synthetic peptide directed towards the C terminal of human Slc26a2. Peptide sequence VSMQLSHDPLEVHTIVIDCSAIQFLDTAGIHTLKEVRRDYEAVGIQVLLA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
D5S1708, Diastrophic dysplasia protein, diastrophic dysplasia sulfate transporter, DTDMSTP157, DTDSTMST153, EDM4, solute carrier family 26 (sulfate transporter), member 2, Solute carrier family 26 member 2, sulfate anion transporter 1, sulfate transporter | |
SLC26A2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title