Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC26A3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096279 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096279 20 μL
NB096278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096279 Supplier Novus Biologicals Supplier No. NBP30507920UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC26A3 Polyclonal specifically detects SLC26A3 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC26A3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias chloride anion exchanger, CLD, congenital chloride diarrhea, down-regulated in adenoma protein, DRADown-regulated in adenoma, Protein DRA, Solute carrier family 26 member 3, solute carrier family 26, member 3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 505-764 of human SLC26A3 (NP_000102.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1811
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.