Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC26A3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | SLC26A3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SLC26A3 Polyclonal specifically detects SLC26A3 in Human samples. It is validated for Western Blot.Specifications
SLC26A3 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
1811 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
chloride anion exchanger, CLD, congenital chloride diarrhea, down-regulated in adenoma protein, DRADown-regulated in adenoma, Protein DRA, Solute carrier family 26 member 3, solute carrier family 26, member 3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 505-764 of human SLC26A3 (NP_000102.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title