Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC28A3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229615SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229615SS 0.025 mg
NBP229615 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229615SS Supplier Novus Biologicals Supplier No. NBP229615SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC28A3 Polyclonal specifically detects SLC28A3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC28A3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 4 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9HAS3
Gene Alias CNT 3, CNT3concentrative Na+-nucleoside cotransporter, Concentrative Na(+)-nucleoside cotransporter 3, hCNT3, solute carrier family 28 (sodium-coupled nucleoside transporter), member 3, solute carrier family 28 member 3
Gene Symbols SLC28A3
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SLC28A3 protein (between residues 30-100) [NCBI NP_071410.1]
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 64078
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.