Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC29A4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241314 0.1 mg
NB435919 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241314 Supplier Novus Biologicals Supplier No. NBP241314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC29A4 Polyclonal specifically detects SLC29A4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC29A4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Alias ENT4PMAT, equilibrative nucleoside transporter 4, FLJ34923, hENT4, Plasma membrane monoamine transporter, solute carrier family 29 (nucleoside transporters), member 4, Solute carrier family 29 member 4
Gene Symbols SLC29A4
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the center of human SLC29A4. The immunogen is located within amino acids 310 - 360 of SLC29A4. Amino Acid Squence: MRFDVPRPRVQRSWPTFR
Molecular Weight of Antigen 58 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222962
Test Specificity SLC29A4 antibody is human specific. SLC29A4 antibody is predicted to not cross-react with other SLC29 proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.