Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC2A13 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317114100UL
This item is not returnable.
View return policy
Description
SLC2A13 Polyclonal antibody specifically detects SLC2A13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC2A13 | |
Polyclonal | |
Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
H(+)-myo-inositol cotransporter, H(+)-myo-inositol symporter, Hmit, HMITproton (H+) myo-inositol symporter, MGC48624, proton myo-inositol cotransporter, solute carrier family 2 (facilitated glucose transporter), member 13 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: PESPRWLIQKGQTQKARRILSQMRGNQTIDEEYDSIKNNIEEEEKEVGSAGPVICRMLSYP | |
100 μg | |
Alzheimers Research, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
114134 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction