Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC2A13 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$366.50 - $609.50
Specifications
Antigen | SLC2A13 |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC2A13 Polyclonal antibody specifically detects SLC2A13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC2A13 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
114134 | |
IgG | |
Affinity purified |
Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
H(+)-myo-inositol cotransporter, H(+)-myo-inositol symporter, Hmit, HMITproton (H+) myo-inositol symporter, MGC48624, proton myo-inositol cotransporter, solute carrier family 2 (facilitated glucose transporter), member 13 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: PESPRWLIQKGQTQKARRILSQMRGNQTIDEEYDSIKNNIEEEEKEVGSAGPVICRMLSYP | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title