Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC30A5 Antibody, Novus Biologicals™
SDP

Catalog No. NB013596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013596 Supplier Novus Biologicals Supplier No. NBP280523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC30A5 Polyclonal specifically detects SLC30A5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC30A5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:2000, Immunohistochemistry-Paraffin 1:2000
Formulation Lyophilized from whole serum
Gene Alias FLJ12756, MGC5499, solute carrier family 30 (zinc transporter), member 5, zinc transporter 5, zinc transporter ZTL1, ZnT-5FLJ12496, ZNT5Solute carrier family 30 member 5, ZNTL1hZTL1, ZTL1ZnT-like transporter 1
Gene Symbols SLC30A5
Host Species Rabbit
Immunogen A synthetic peptide from internal cytoplasmic region of human SLC30A5 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and mouse.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64924
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.