Learn More
Description
Specifications
Specifications
| Antigen | SLC31A1/CTR1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 5E6.1B4 |
| Conjugate | Biotin |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | O15431 |
| Gene Alias | Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 |
| Gene Symbols | SLC31A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
