Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC31A1/CTR1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB100402G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100402G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100402G Supplier Novus Biologicals Supplier No. NB100402G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC31A1/CTR1 Polyclonal specifically detects SLC31A1/CTR1 in Canine, Human, Mouse, Pig, Rat, Xenopus species, Zebrafish samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen SLC31A1/CTR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O15431
Gene Alias Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1
Gene Symbols SLC31A1
Host Species Rabbit
Immunogen A synthetic peptide derived from a C-terminal sequence of human SLC31A1/CTR1 [Uniprot: O15431]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1317
Target Species Canine, Human, Mouse, Pig, Rat, Xenopus species, Zebrafish
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.