Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC31A1/CTR1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100402SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100402SS 0.025 mL
NB100402 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100402SS Supplier Novus Biologicals Supplier No. NB100402SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 24 publications

SLC31A1/CTR1 Polyclonal specifically detects SLC31A1/CTR1 in Canine, Human, Mouse, Pig, Rat, Xenopus species, Zebrafish samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen SLC31A1/CTR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen
Formulation Tris-Citrate/Phosphate (pH 7 - 8) with 0.1% Sodium Azide
Gene Accession No. O15431
Gene Alias Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1
Gene Symbols SLC31A1
Host Species Rabbit
Immunogen A synthetic peptide derived from a C-terminal sequence of human SLC31A1/CTR1 [Uniprot: O15431]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1317
Target Species Canine, Mouse, Human, Mouse, Pig, Zebrafish, Rat, Xenopus species, Zebrafish
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.