Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC31A1/CTR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 24 publications
$192.47 - $493.10
Specifications
| Antigen | SLC31A1/CTR1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100402SS
![]() |
Novus Biologicals
NB100402SS |
0.025 mL |
Each for $192.47
|
|
|||||
NB100402
![]() |
Novus Biologicals
NB100402 |
0.1 mL |
Each for $493.10
|
|
|||||
Description
SLC31A1/CTR1 Polyclonal specifically detects SLC31A1/CTR1 in Human, Mouse, Rat, Porcine, Canine, Xenopus, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SLC31A1/CTR1 | |
| Western Blot, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Rabbit | |
| Neuroscience, Neurotransmission, Protein Kinase | |
| Tris-Citrate/Phosphate (pH 7 - 8) with 0.1% Sodium Azide | |
| Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 | |
| SLC31A1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Xenopus species, Zebrafish | |
| O15431 | |
| 1317 | |
| A synthetic peptide derived from a C-terminal sequence of human SLC31A1/CTR1 [Uniprot: O15431] | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title