Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC31A1/CTR1 Rabbit anti-Human, Mouse, Rat, Clone: 4C10C5, Novus Biologicals™
SDP

Catalog No. NB165705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165705 20 μg
NB165704 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165705 Supplier Novus Biologicals Supplier No. NBP31530420UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen SLC31A1/CTR1
Applications Western Blot
Classification Monoclonal
Clone 4C10C5
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC31A1/CTR1 (O15431). MDHSHHMGMSYMDSNSTMQPSHHHPTTSASHSHGGGDSSMMMMPMTFYFGFKNVELLFSGLVINTAGEMAGAFVAVFLLAMFYEGLKIARESLLRKSQVS
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1317
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.