Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC34A1 Antibody (10B1.3E9), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N242216P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N242216P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N242216P594 Supplier Novus Biologicals Supplier No. NBP242216PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC34A1 Monoclonal specifically detects SLC34A1 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen SLC34A1
Applications Flow Cytometry
Classification Monoclonal
Clone 10B1.3E9
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FRTS2, Na(+)/Pi cotransporter 2A, Na(+)-dependent phosphate cotransporter 2A, NaPi-2a, NaPi-3, NPT2NPHLOP1, NPTIIa, renal sodium-dependent phosphate transporter, SLC11, SLC17A2Na+-phosphate cotransporter type II, sodium/phosphate co-transporter, Sodium/phosphate cotransporter 2A, sodium-dependent phosphate transport protein 2A, Sodium-phosphate transport protein 2A, solute carrier family 17 (sodium phosphate), member 2, solute carrier family 34 (sodium phosphate), member 1, Solute carrier family 34 member 1
Gene Symbols SLC34A1
Host Species Mouse
Immunogen Two synthetic peptides made to an internal portions of the human SLC34A1 protein (between amino acids 25-150 and 100-200) [UniProt Q06495]
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6569.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.