Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC34A3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB136673 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136673 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136673 Supplier Novus Biologicals Supplier No. NBP306366JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC34A3 Polyclonal specifically detects SLC34A3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC34A3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Na(+)/Pi cotransporter 2C, Na(+)-dependent phosphate cotransporter 2C, naPi-2c, sodium-phosphate transport protein 2C, solute carrier family 34 (sodium phosphate), member 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC34A3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142680
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.