Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35B3 Antibody, Novus Biologicals™
SDP

Catalog No. NB436363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436363 25 μL
NBP237943 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436363 Supplier Novus Biologicals Supplier No. NBP23794325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35B3 Polyclonal specifically detects SLC35B3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC35B3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9H1N7
Gene Alias 3' phosphoadenosine 5' phosphosulfate transporter 2, adenosine 3'-phospho 5'-phosphosulfate transporter 2, C6orf196, CGI-19, chromosome 6 open reading frame 196, PAPS transporter 2, PAPST2dJ453H5.1,3'-phosphoadenosine 5'-phosphosulfate transporter, Solute carrier family 35 member B3, solute carrier family 35, member B3
Gene Symbols SLC35B3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: VIGNVQEKAMKLHNASNSEMVLY
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51000
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.