Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35D2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB031479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB031479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB031479 Supplier Novus Biologicals Supplier No. NBP2835500.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35D2 Polyclonal specifically detects SLC35D2 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC35D2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias HFRC, HFRC1UDP-N-acetylglucosamine transporter, Homolog of Fringe connection protein 1, MGC117215, MGC142139, Solute carrier family 35 member D2, solute carrier family 35, member D2, SQV7Lfringe connection, SQV7-like protein, UDP-galactose transporter-related protein 8, UDP-N-acetylglucosamine/UDP-glucose/GDP-mannose transporter, UGTREL8, UGTrel8hfrc
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Human SLC35D2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11046
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.