Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35D3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342191 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342191 Supplier Novus Biologicals Supplier No. NBP281839MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35D3 Polyclonal antibody specifically detects SLC35D3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SLC35D3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bA55K22.3, frc, fringe-like 1, FRCL1, Fringe connection-like protein 1, MGC102873, solute carrier family 35 member D3, solute carrier family 35, member D3
Host Species Rabbit
Immunogen SLC35D3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3. The immunogen is located within the last 50 amino acids of SLC35D3. Amino Acid Squence: LVRGTRYMKKDYLIENE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 340146
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.