Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35E1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB329980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329980 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329980 Supplier Novus Biologicals Supplier No. NBP297796F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35E1 Polyclonal antibody specifically detects SLC35E1 in Human samples. It is validated for Western Blot

Specifications

Antigen SLC35E1
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias FLJ14251, FLJ36689, solute carrier family 35 member E1, solute carrier family 35, member E1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC35E1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79939
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.