Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC36A4 Antibody (PAT-4/9/H10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20237A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20237A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20237A32 Supplier Novus Biologicals Supplier No. NBP250237AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC36A4 Monoclonal specifically detects SLC36A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockdown Validated.

Specifications

Antigen SLC36A4
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Monoclonal
Clone PAT-4/9/H10
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias proton/amino acid transporter 4, solute carrier family 36 (proton/amino acid symporter), member 4
Gene Symbols SLC36A4
Host Species Mouse
Immunogen Antigenic amino acid sequence within the N-terminus of SLC36A4 (REELDMDVMRPLINE-C).
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 120103
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.