Learn More
Description
Specifications
Specifications
| Antigen | SLC38A1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | S104-32 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Amino acid transporter A1, ATA1SNAT1, NAT2N-system amino acid transporter 2, SAT1amino acid transporter system A1, sodium-coupled neutral amino acid transporter 1, Solute carrier family 38 member 1, solute carrier family 38, member 1, System A amino acid transporter 1, System N amino acid transporter 1 |
| Gene Symbols | SLC38A1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
