Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC38A4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155228 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155228 Supplier Novus Biologicals Supplier No. NBP155228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC38A4 Polyclonal specifically detects SLC38A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC38A4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q969I6
Gene Alias Amino acid transporter A3, ATA3MGC126876, FLJ10191, N amino acid transporter 3, Na(+)-coupled neutral amino acid transporter 4, NAT3amino acid transporter system A3, PAAT, SNAT4, sodium-coupled neutral amino acid transporter 4, Solute carrier family 38 member 4, solute carrier family 38, member 4, System A amino acid transporter 3, System N amino acid transporter 3
Gene Symbols SLC38A4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC38A4(solute carrier family 38, member 4) The peptide sequence was selected from the middle region of SLC38A4. Peptide sequence LAALFGYLTFYGEVEDELLHAYSKVYTLDIPLLMVRLAVLVAVTLTVPIV.
Molecular Weight of Antigen 17 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55089
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.