Learn More
Invitrogen™ SLC38A9 Polyclonal Antibody

Description
SLC38A9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Specifications
Specifications
| Antigen | SLC38A9 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | SLC38A9 |
| Gene Accession No. | Q8NBW4 |
| Gene Alias | 4833412L08Rik; 6720411P22Rik; 9130023D20Rik; 9430067K09Rik; A730092C09; putative sodium-coupled neutral amino acid transporter 9; RGD1311881; Slc38a9; Sodium-coupled neutral amino acid transporter 9; solute carrier family 38 member 9; solute carrier family 38, member 9; up-regulated in lung cancer 11; URLC11 |
| Gene Symbols | SLC38A9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.