Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC39A2/ZIP2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176499 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176499 Supplier Novus Biologicals Supplier No. NBP176499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC39A2/ZIP2 Polyclonal specifically detects SLC39A2/ZIP2 in Human, Mouse, Rat, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC39A2/ZIP2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_055394
Gene Alias Eti-1, MGC119190, solute carrier family 39 (zinc transporter), member 2, Solute carrier family 39 member 2, zinc transporter ZIP2, ZIP-2, ZIP2hZIP2, Zrt- and Irt-like protein 2,6A1
Gene Symbols SLC39A2
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZIP2. The immunogen is located within amino acids 70 - 120 of ZIP2. Amino Acid Squence: NRSASERNSSGDADSAH
Molecular Weight of Antigen 32 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29986
Target Species Human, Mouse, Rat, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.