Learn More
Description
Specifications
Specifications
| Antigen | SLC39A5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | LZT-Hs7, MGC34778, solute carrier family 39 (metal ion transporter), member 5, Solute carrier family 39 member 5, zinc transporter ZIP5, ZIP5, ZIP-5, Zrt- and Irt-like protein 5 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 305-385 of human SLC39A5 (NP_775867.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
