Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC39A5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | SLC39A5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SLC39A5 Polyclonal specifically detects SLC39A5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SLC39A5 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
283375 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
LZT-Hs7, MGC34778, solute carrier family 39 (metal ion transporter), member 5, Solute carrier family 39 member 5, zinc transporter ZIP5, ZIP5, ZIP-5, Zrt- and Irt-like protein 5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 305-385 of human SLC39A5 (NP_775867.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title