Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC39A8/ZIP8 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176505 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176505 Supplier Novus Biologicals Supplier No. NBP176505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC39A8/ZIP8 Polyclonal specifically detects SLC39A8/ZIP8 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC39A8/ZIP8
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_071437
Gene Alias BCG induced integral membrane protein BIGM103, BIGM103BCG-induced integral membrane protein in monocyte clone 103 protein, LIV-1 subfamily of ZIP zinc transporter 6, LZT-Hs6, solute carrier family 39 (metal ion transporter), member 8, solute carrier family 39 (zinc transporter), member 8, Solute carrier family 39 member 8, zinc transporter ZIP8, ZIP8, ZIP-8, Zrt- and Irt-like protein 8
Gene Symbols SLC39A8
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human ZIP8. The immunogen is located within the last 50 amino acids of ZIP8. Amino Acid Squence: PEMDMLREKVTGRKTDF
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64116
Test Specificity ZIP8 antibody is predicted to not cross-react with other ZIP family members.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.