Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC39A8/ZIP8 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB050725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050725 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050725 Supplier Novus Biologicals Supplier No. NBP297997UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC39A8/ZIP8 Polyclonal specifically detects SLC39A8/ZIP8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC39A8/ZIP8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BCG induced integral membrane protein BIGM103, BIGM103BCG-induced integral membrane protein in monocyte clone 103 protein, LIV-1 subfamily of ZIP zinc transporter 6, LZT-Hs6, solute carrier family 39 (metal ion transporter), member 8, solute carrier family 39 (zinc transporter), member 8, Solute carrier family 39 member 8, zinc transporter ZIP8, ZIP8, ZIP-8, Zrt- and Irt-like protein 8
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC39A8/ZIP8 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64116
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.