Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC3A1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB335337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335337 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335337 Supplier Novus Biologicals Supplier No. NBP298113APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC3A1 Polyclonal antibody specifically detects SLC3A1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SLC3A1
Applications Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias activator of cystine, dibasic and neutral amino acid transport), member 1, amino acid transporter 1, ATR1SLC3A1 variant C, B(0+)-type amino acid transport protein, CSNU1, D2h, D2HSLC3A1 variant D, dibasic and neutral amino acid transporters), FLJ34681, member 1, NBATneutral and basic amino acid transport protein rBAT, RBATSLC3A1 variant B, SLC3A1 variant E, SLC3A1 variant F, SLC3A1 variant G, solute carrier family 3 (cystine, dibasic and neutral amino acid transporters
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC3A1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6519
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.