Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC3A1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB052598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052598 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052598 Supplier Novus Biologicals Supplier No. NBP298113G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC3A1 Polyclonal specifically detects SLC3A1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SLC3A1
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias activator of cystine, dibasic and neutral amino acid transport), member 1, amino acid transporter 1, ATR1SLC3A1 variant C, B(0+)-type amino acid transport protein, CSNU1, D2h, D2HSLC3A1 variant D, dibasic and neutral amino acid transporters), FLJ34681, member 1, NBATneutral and basic amino acid transport protein rBAT, RBATSLC3A1 variant B, SLC3A1 variant E, SLC3A1 variant F, SLC3A1 variant G, solute carrier family 3 (cystine, dibasic and neutral amino acid transporters
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC3A1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6519
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.