Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC40A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986215
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6215 Supplier Bioss Supplier No. BS4906R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.

Specifications

Antigen SLC40A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC40A1
Gene Accession No. Q9NP59
Gene Alias CAR1; Cell adhesion regulator; duodenal-specific; Dusg; Ferroportin; ferroportin 1; Ferroportin-1; FPN1; Fpt1; HFE4; Ireg1; iron regulated gene 1; Iron-regulated transporter 1; Metal transporter protein 1; metal transporting protein 1; MST079; MSTP079; MTP; MTP1; Ol5; Pcm; polycythaemia; Slc11a3; SLC11A3 iron transporter; Slc39a1; Slc40a1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 39 (iron-regulated transporter), member 1; solute carrier family 40 (iron-regulated transporter), member 1; Solute carrier family 40 member 1
Gene Symbols SLC40A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC40A1/FPN1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30061
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.