Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC45A3/Prostein Antibody, Novus Biologicals™
SDP

Catalog No. NB405885 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB405885 25 μL
NBP189630 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB405885 Supplier Novus Biologicals Supplier No. NBP18963025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

SLC45A3/Prostein Polyclonal specifically detects SLC45A3/Prostein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC45A3/Prostein
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Western Blot Reported in scientific literature (PMID: 30719129)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3
Gene Symbols SLC45A3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:AEEAALGPTEPAEGLSAPSLSPHCCPCRARLAFRNLGALLPRLHQLCCRMP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85414
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.