Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC45A3/Prostein Antibody (SLC45A3/7176R), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB187155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187155 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187155 Supplier Novus Biologicals Supplier No. NBP314049UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SLC45A3/Prostein Monoclonal antibody specifically detects SLC45A3/Prostein in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SLC45A3/Prostein
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SLC45A3/7176R
Conjugate DyLight 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3
Host Species Rabbit
Immunogen Synthesized peptides to the N terminus of human SLC45A3/Prostein protein (Uniprot: Q96JT2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85414
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.