Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC45A3/Prostein Rabbit anti-Human, Clone: RM426, Novus Biologicals™
SDP

Catalog No. NB119703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119703 100 μg
NB119704 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB119703 Supplier Novus Biologicals Supplier No. NBP309178100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SLC45A3/Prostein Monoclonal specifically detects SLC45A3/Prostein in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen SLC45A3/Prostein
Applications Immunohistochemistry
Classification Monoclonal
Clone RM426
Conjugate Unconjugated
Dilution Immunohistochemistry 1:100-1:400
Formulation PBS, 50% Glycerol, 1% BSA
Gene Alias IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3
Host Species Rabbit
Immunogen A peptide corresponding to the C-terminus of human SLC45A3/Prostein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85414
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.