Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC45A3/Prostein Rabbit anti-Human, Clone: RM426, Novus Biologicals™

Rabbit Monoclonal Antibody
$181.90 - $548.00
Specifications
Antigen | SLC45A3/Prostein |
---|---|
Clone | RM426 |
Dilution | Immunohistochemistry 1:100-1:400 |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Description
SLC45A3/Prostein Monoclonal specifically detects SLC45A3/Prostein in Human samples. It is validated for Immunohistochemistry.Specifications
SLC45A3/Prostein | |
Immunohistochemistry 1:100-1:400 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 50% Glycerol, 1% BSA | |
85414 | |
IgG | |
Protein A purified |
RM426 | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3 | |
A peptide corresponding to the C-terminus of human SLC45A3/Prostein | |
Primary | |
Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title