Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC45A4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB319946 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319946 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319946 Supplier Novus Biologicals Supplier No. NBP298631MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC45A4 Polyclonal antibody specifically detects SLC45A4 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SLC45A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias KIAA1126, solute carrier family 45, member 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC45A4 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57210
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.