Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A11 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15963520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15963520 20 μL
NBP159635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15963520 Supplier Novus Biologicals Supplier No. NBP15963520UL

Rabbit Polyclonal Antibody

SLC5A11 Polyclonal specifically detects SLC5A11 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC5A11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8WWX8
Gene Alias KST1RKST1, Na(+)/myo-inositol cotransporter 2, na+/myo-inositol cotransporter 2, putative sodium-coupled cotransporter RKST1, SGLT6, SLGTX, SMIT2homolog of rabbit KST1, Sodium/glucose cotransporter KST1, sodium/myo-inositol cotransporter 2, Sodium-dependent glucose cotransporter, solute carrier family 5 (sodium/glucose cotransporter), member 11, Solute carrier family 5 member 11
Gene Symbols SLC5A11
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC5A11(solute carrier family 5 (sodium/glucose cotransporter), member 11) The peptide sequence was selected from the middle region of SLC5A11. Peptide sequence VGGMEGLKEKYFLALASNRSENSSCGLPREDAFHIFRDPLTS
Molecular Weight of Antigen 74 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115584
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.