Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11040431 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040431 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040431 Supplier Novus Biologicals Supplier No. NB11040431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC5A3 Polyclonal specifically detects SLC5A3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SGLT1/SLC5A1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 20 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias D22S675, High affinity sodium-glucose cotransporter, Na+/glucose cotransporter 1, NAGTsodium/glucose cotransporter 1, SGLT1Na(+)/glucose cotransporter 1, solute carrier family 5 (sodium/glucose cotransporter), member 1, Solute carrier family 5 member 1
Gene Symbols SLC5A1
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminus of human SLC5A3.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 6523
Test Specificity This is specific to the human SLC5A3 protein. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human, Pig, Bat, Bovine, Canine, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.