Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A5/Sodium Iodide Symporter Antibody (14F), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP259341C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259341C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259341C Supplier Novus Biologicals Supplier No. NBP259341C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SLC5A5/Sodium Iodide Symporter
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 14F
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Gene Symbols SLC5A5
Host Species Mouse
Immunogen Mannose binding protein hNIS fusion (AA468-643)
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 6528
Test Specificity Detects 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at 30 kDa, and 15kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.