Learn More
SLC5A5/Sodium Iodide Symporter Antibody (14F), FITC, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | SLC5A5/Sodium Iodide Symporter |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 14F |
| Conjugate | FITC |
| Dilution | Immunohistochemistry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 |
| Gene Symbols | SLC5A5 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.