Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A5/Sodium Iodide Symporter Antibody (902320), HRP, Novus Biologicals™
SDP

Catalog No. NB346163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346163 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346163 Supplier Novus Biologicals Supplier No. FAB8367H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen SLC5A5/Sodium Iodide Symporter
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 902320
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC5A5, Accession # Q92911
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6528
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.