Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A5/Sodium Iodide Symporter Antibody (FP5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP170342UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP170342UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP170342UV Supplier Novus Biologicals Supplier No. NBP170342UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC5A5/Sodium Iodide Symporter
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FP5
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Gene Symbols SLC5A5
Host Species Mouse
Immunogen Mannose binding protein hNIS fusion (AA468-643)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6528
Test Specificity Detects approx 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at approx 30 kDa, and approx 15kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.