Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A8/SMCT1 Antibody (903502), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346121 Supplier Novus Biologicals Supplier No. FAB8398MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC5A8/SMCT1 Monoclonal specifically detects SLC5A8/SMCT1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen SLC5A8/SMCT1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 903502
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AITSolute carrier family 5 member 8, Apical iodide transporter, Electrogenic sodium monocarboxylate cotransporter, MGC125354, SMCTSMCT1, Sodium iodide-related cotransporter, sodium-coupled monocarboxylate transporter 1, solute carrier family 5 (iodide transporter), member 8
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC5A8/SMCT1, Met1-Leu610, Accession # NP_666018
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160728
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.