Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A9 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247137 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247137 Supplier Novus Biologicals Supplier No. NBP247137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC5A9 Polyclonal specifically detects SLC5A9 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC5A9
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias hSGLT4, MGC132523, Na(+)/glucose cotransporter 4, SGLT4MGC132517, sodium/glucose cotransporter 4, solute carrier family 5 (sodium/glucose cotransporter), member 9, Solute carrier family 5 member 9
Gene Symbols SLC5A9
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from C-terminus of human SLC5A9.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200010
Test Specificity This is specific to the human SLC5A9 protein. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.