Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A12 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226172 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226172 Supplier Novus Biologicals Supplier No. NBP226172
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SLC6A12 Polyclonal specifically detects SLC6A12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.

Specifications

Antigen SLC6A12
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias BGT1, FLJ38727, gamma-aminobutyric acid transporter, solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12
Gene Symbols SLC6A12
Host Species Goat
Immunogen Peptide with sequence CLDGSAGRNFGPSPT corresponding to C-Terminus according to NP_003035.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6539
Test Specificity Reported variants represent identical protein: NP_001193860.1, NP_003035.3, NP_001116319.1, NP_001116320.1.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.