Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A18 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB043759 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043759 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043759 Supplier Novus Biologicals Supplier No. NBP297457F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC6A18 Polyclonal specifically detects SLC6A18 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC6A18
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ31236, Sodium- and chloride-dependent transporter XTRP2, sodium channel-like protein, sodium-dependent neutral amino acid transporter B(0)AT3, solute carrier family 6 (neurotransmitter transporter), member 18, Solute carrier family 6 member 18, solute carrier family 6, member 18, System B(0) neutral amino acid transporter AT3, XTRP2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC6A18 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348932
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.