Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A18 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB331650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331650 Supplier Novus Biologicals Supplier No. NBP297457MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC6A18 Polyclonal antibody specifically detects SLC6A18 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SLC6A18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ31236, Sodium- and chloride-dependent transporter XTRP2, sodium channel-like protein, sodium-dependent neutral amino acid transporter B(0)AT3, solute carrier family 6 (neurotransmitter transporter), member 18, Solute carrier family 6 member 18, solute carrier family 6, member 18, System B(0) neutral amino acid transporter AT3, XTRP2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC6A18 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 348932
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.